Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lightning-Link (R) Rapid Alexa Fluor 568 Antibody Labeling Kit
SDP

Catalog No. 3340030 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-400-30 30 μg
33-400-10 300 μg
33-400-15 1 mg
33-400-05 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 33-400-30 Supplier R&D Systems™ Supplier No. 3340030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Antibody labeling kit

  • Simple conjugation workflow with minimal hands-on time.
  • Requires purified antibody in a compatible buffer.
  • Conjugated antibodies can be used without further purification.

Specifications

Quantity 30 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C
Label Type Alexa Fluor
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.