Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lightning-Link (R) Rapid Atto488 Antibody Labeling Kit
SDP

Catalog No. 3500030 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-000-30 30 μg
35-000-10 300 μg
35-000-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 35-000-30 Supplier R&D Systems™ Supplier No. 3500030

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Applications: Flow Cytometry

Atto488 is one of a new generation of fluorescent labels, which has been optimised for excitation with an argon laser. It has a strong absorption at 501nm, high fluorescence at 523nm (extinction coefficient 9.0 x10 cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

Specifications

Product Type Rapid Atto488 Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate Atto 488
Product Line Novus
Quantity 30 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.