Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Atto633 Antibody Labeling Kit
SDP

Catalog No. 3530010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-300-10 300 μg
35-300-30 30 μg
35-300-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 35-300-10 Supplier Novus Biologicals™ Supplier No. 3530010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody labeling kit

Atto633 is one of a new generation of fluorescent labels. It has a strong absorption at 629nm, high fluorescence at 657nm (extinction coefficient 1.3 x10&superscript_begin;5&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield. Atto633 can be used as a substitute dye for DY-633, LightCycler Red 640, Alexa Fluor 633, and Cy 5.

Order Info

Quantity

Specifications

Product Type Rapid Atto633 Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate Atto 633
Quantity 300 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.