Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Cy5 Antibody Labeling Kit
SDP

Catalog No. p-7070016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-200-30 30 μg
34-200-10 300 μg
34-200-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 34-200-30 Supplier Novus Biologicals™ Supplier No. 3420030
Only null left
Add to Cart
Add to Cart

Antibody labeling kit

Cyanine Dye 5 (Cy5) has a strong absorption peak at 643nm and a secondary absorption peak at 605nm. The maximum emission peak is at 667nm, with an excitation coefficient of 2.5 x10&superscript_begin;5&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;. Cy5 can be used as an alternative to AlexaFluor 647, and the phycobiliprotein Allophycocyanin.

Order Info

Quantity

Specifications

Conjugate Cy5
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Product Type Rapid Cy5 Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.