Learn More
R&D Systems™ Lightning-Link (R) Rapid DyLight 350 Antibody Labeling Kit
SDP
Catalog No. NB092258 Shop All Novus Products
Change view
Click to view available options
Quantity:
300 μg
Catalog No. Quantity
NB092258 300 μg
1 options
Catalog No. NB092258 Supplier R&D Systems™ Supplier No. 3200010300UG
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. The easy-to-use, one step procedure allows researchers to covalently label biomolecules with only 30 seconds hands-on time; conjugates are ready to use in less than twenty minutes
Specifications
Content And Storage -20°C
Product Line Novus
Quantity 300 μg