Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lightning-Link (R) Rapid DyLight 405 Antibody Labeling Kit
SDP

Catalog No. 3210015 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
300 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-100-15 1 mg
32-100-10 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-100-15 Supplier R&D Systems™ Supplier No. 3210015

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Antibody labeling kit

DyLight 405 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 400nm, high fluorescence at 420nm (extinction coefficient 3.0 x10&superscript_begin;4&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.

Order Info

Quantity

Specifications

Product Type Rapid DyLight 405 Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Conjugate DyLight 405
Product Line Novus
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.