Description
- Simple conjugation workflow with minimal hands-on time.
- Requires purified antibody in a compatible buffer.
- Conjugated antibodies can be used without further purification.
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | DyLight 488 Antibody Labeling Kit |
| Label Type | DyLight Fluors |
| Product Line | Novus |