Learn More
Description
- Lightning-Link™ Rapid technology can be used to label small quantities of antibody with 100% recovery
- Easy to use antibody labeling kit with quick labeling times
- From 10μg up to 2mg
- Kit with 30 seconds hands-on time
- No separation steps and no losses
- Scalable technology
- Western Blotting, ELISA, Immunohistochemistry, Immunofluorescence and FACS analysis
Order Info
Quantity
Specifications
Specifications
| Product Type | Rapid DyLight 633 Antibody Labeling Kit |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Conjugate | DyLight 633 |
| Product Line | Novus |
| Quantity | 300 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
