Learn More
Description
- Simple conjugation workflow with minimal hands-on time.
- Requires purified antibody in a compatible buffer.
- Conjugated antibodies can be used without further purification.
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | Antibody Labeling Kit |
| Content And Storage | -20°C |
| Label Type | DyLight Fluors |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
