Learn More
Novus Biologicals™ Lightning-Link (R) Rapid DyLight 680 Antibody Labeling Kit
SDP
Catalog No. 3270005 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
32-700-05 100 μg
1 options
Catalog No. 32-700-05 Supplier Novus Biologicals™ Supplier No. 3270005
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
Specifications
Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C