Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lightning-Link (R) Rapid DyLight 755 Antibody Labeling Kit
SDP

Catalog No. 3280005 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-800-05 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-800-05 Supplier R&D Systems™ Supplier No. 3280005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lightning-Link Rapid DyLight 755 Antibody Labeling Kits enable fluorescent labeling of purified antibodies.

  • Simple conjugation workflow with minimal hands-on time.
  • Requires purified antibody in a compatible buffer.
  • Conjugated antibodies can be used without further purification.

Specifications

Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C
Label Type DyLight Fluors
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.