Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lightning-Link (R) Rapid Rhodamine Antibody Labeling Kit
SDP

Catalog No. 3110010 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
30 μg
300 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-100-10 300 μg
31-100-15 1 mg
31-100-30 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For Research Use Only
Catalog No. 31-100-10 Supplier R&D Systems™ Supplier No. 3110010

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

For Research Use Only

A Novus product, part of R&D Systems. Label antibodies, proteins, peptides and other biomolecules with rhodamine, using simple procedure. Set up conjugations in seconds, and use them within minutes.

  • No desalting or dialysis steps
  • Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Rhodamine used in kit has absorption peaks at 520 and 544nm
  • Maximum emission is obtained when the rhodamine is excited at 544nm

Specifications

Product Type Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 300 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.