Learn More
Description
- The easy-to-use, one step procedure allows researchers to covalently label biomolecules with only 30 seconds hands-on time.
- Conjugates are ready to use in less than twenty minutes.
- Simply pipettes the biomolecule into a vial of lyophilized mixture containing the label of interest and incubates (for more details please watch the video below)
- Texas red is a red fluorescent dye, used in histology, in fluorescence microscopy applications and in immunohistochemistry
- It has a strong absorption at 596nm, with a high fluorescence at 616nm (extinction coefficient 8.0 x 104cm-1M-1) and high quantum yield
- By circumventing the desalting or dialysis steps that commonly interrupt traditional antibody conjugation procedures, LightningLink technology can be used to label both small and large quantities of primary antibodies with ease
- Batch-to-batch variation upon scale up is minimal as the process is so simple, and recoveries are always 100%
- Includes: 1 vial of LL Rapid Modifier reagent, 1 vial of LL Rapid Quencher reagent, 1 glas cial of LIghtning Link Rapid mix
Specifications
Specifications
| Content And Storage | -20°C |
| Product Line | Novus |
| Quantity | 100 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
