Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Texas Red Antibody Labeling Kit
Shop All Novus Biologicals Products

Description
Order Info
Quantity
Specifications
Specifications
| Product Type | Rapid Texas Red Antibody Labeling Kit |
| Content And Storage | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent. Store at -20°C. Avoid freeze-thaw cycles. |
| Conjugate | Texas Red |
| Quantity | 30 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.