Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Rhodamine Antibody Labeling Kit
Shop All Novus Biologicals Products

Description
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
- Lightning-Link™ Rapid mix
- 1 vial of LL Rapid Quencher reagent
- 1 vial of LL Rapid Modifier reagent
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | Antibody Labeling Kit |
| Content And Storage | -20°C |
| Target | Lightning-Link (R) Rapid Rhodamine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.