Learn More
R&D Systems™ Lightning-Link (R) Streptavidin Antibody Labeling Kit
SDP
Catalog No. 7080005 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
70-800-05 100 μg
1 options
Catalog No. 70-800-05 Supplier R&D Systems™ Supplier No. 7080005
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems. R&D Systems™ Lightning-Link Streptavidin Antibody Labeling Kit enables the direct conjugation of streptavidin to any biomolecule with an available amine group.
  • Enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in research and development applications, drug discovery and the development of diagnostic kits.
  • Researcher simply pipettes their antibody or other biomolecule into the vial of Lightning-Link Streptavidin and incubates for 3 hours.
  • Streptavidin is a 53kDa protein purified from Streptomyces avidinii, which has widespread applications due to its very high affinity to the vitamin biotin. Streptavidin is a tetrameric molecule, composed of 4 13kDa monomers, each of which can bind a molecule of biotin.
  • The kit is unique in the sense that it is the only DIY streptavidin labeling kit available on the market. Like our biotinylation kits, Lightning-Link Streptavidin has been optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step. The way in which the kits are optimised means the antibody or protein of interest will be labeled with up to 3 streptavidin molecules each, maximising the opportunity for biotin to bind and create a biotin streptavidin complex.
Specifications
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Line Novus