Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Lightweight Pigskin Leather MIG Welder's Gloves

Catalog No. 19840313
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-840-313 X-Large
19-840-311 Medium
19-840-312 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-840-313 Supplier Black Stallion™ Supplier No. 26XL
This item is not returnable. View return policy

Lightweight pigskin leather provides supple protection in a classic design for MIG welding applications.

  • Lightweight grain pigskin leather provides supple protection
  • Stitched with Aramid Fiber for additional heat protection
  • Gloves are unlined to increase dexterity
  • Seamless index finger enhances fit and feel
  • Specifically designed for MIG welding applications

Specifications

Color Natural
Hand Specific
Material Grain Pigskin Leather
Size X-Large
For Use With (Application) MIG Welding
Glove Cuff Color Orange
Glove Cuff Style Gauntlet
Series 26
Length (English) Cuff 4.5 in.
Length (Metric) Cuff 11.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.