Learn More
Tecan DiTi Box 1000L for Ref
Supplier: Tecan 30058507
Disposable Tip (DiTi) Box, empty for use with 1000 L refill insert tips, ANSI Format. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. For use with the Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). Set of 10.