Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lilolidine, 98%, Thermo Scientific™

Catalog No. AC452900010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452900010 1 g
AC452900050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452900010 Supplier Thermo Scientific Chemicals Supplier No. 452900010
Add to Cart

Chemical Identifiers

CAS 5840-01-7
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.21
InChI Key QCCKSFHMARIKSK-UHFFFAOYSA-N
IUPAC Name 2-methoxy-4-N-phenylbenzene-1,4-diamine

Specifications

Loss on Drying 1% max. (60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 3-methoxy-n1-phenylbenzene-1,4-diamine, 2-methoxy-n4-phenyl-1,4-phenylenediamine, 4-amino-3-methoxydiphenylamine, 3-methoxy-1-n-phenylbenzene-1,4-diamine, acmc-1apzp, 4-anilino-2-methoxyaniline, p-methoxy-p-aminodiphenylamine, 1,4-benzenediamine, 2-methoxy-n4-phenyl, 2-methoxy-n∼4∼-phenylbenzene-1,4-diamine, 2-methoxy-n4-phenyl-1,4-phenylene diamine
SMILES C1CN2C=CC3=CC=CC(C1)=C23
PubChem CID 611802
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material Lilolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.