Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LILRB4 Recombinant Protein

Catalog No. 89012126 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-012-126 25 μg
89-012-125 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-012-126 Supplier Abnova™ Supplier No. H00011006P0125
Add to Cart
Add to Cart

Human LILRB4 full-length ORF recombinant protein with GST-tag at N-terminal

This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene.

  • Theoretical MW (kDa): 75.02
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MIPTFTALLCLGLSLGPRTDMQAGPLPKPTLWAEPGSVISWGNSVTIWCQGTLEAREYRLDKEESPAPWDRQNPLEPKNKARFSIPSMTEDYAGRYRCYYRSPVGWSQPSDPLELVMTGAYSKPTLSALPSPLVTSGKSVTLLCQSRSPMDTFLLIKERAAHPLLHLRSEHGAQQHQAEFPMSPVTSVHGGTYRCFSSHGFSHYLLSHPSDPLELIVSGSLEGPRPSPTRSVSTAAGPEDQPLMPTGSVPHSGLRRHWEVLIGVLVVSILLLSLLLFLLLQHWRQGKHRTLAQRQADFQRPPGAAEPEPKDGGLQRRSSPAADVQGENFCAAVKNTQPEDGVEMDTRQSPHDEDPQAVTYAKVKHSRPRREMASPPSPLSGEFLDTKDRQAEEDRQMDTEAAASEAPQDVTYARLHSFTLRQKATEPPPSQEGASPAEPSVYATLAIH

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH26309
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 11006
Molecular Weight (g/mol) 75.02
Name LILRB4 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Source Wheat Germ (in vitro)
Immunogen MIPTFTALLCLGLSLGPRTDMQAGPLPKPTLWAEPGSVISWGNSVTIWCQGTLEAREYRLDKEESPAPWDRQNPLEPKNKARFSIPSMTEDYAGRYRCYYRSPVGWSQPSDPLELVMTGAYSKPTLSALPSPLVTSGKSVTLLCQSRSPMDTFLLIKERAAHPLLHLRSEHGAQQHQAEFPMSPVTSVHGGTYRCFSSHGFSHYLLSHPSDPLELIVSGSLEGPRPSPTRSVSTAAGPEDQPLMPTGSVPHSGLRRHWEVLIGVLVVSILLLSLLLFLLLQHWRQGKHRTLAQRQADFQRPPGAAEPEPKDGGLQRRSSPAADVQGENFCAAVKNTQPEDGVEMDTRQSPHDEDPQAVTYAKVKHSRPRREMASPPSPLSGEFLDTKDRQAEEDRQMDTEAAASEAPQDVTYARLHSFTLRQKATEPPPSQEGASPAEPSVYATLAIH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CD85K/HM18/ILT3/LILRB5/LIR-5/LIR5
Common Name LILRB4
Gene Symbol LILRB4
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.