Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Seed Co. Inc Lima Beans, PK
SCI_ED

Catalog No. S000940 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Sufficient For:
Class of 15 students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S000940 Class of 15 students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000940 Supplier American Seed Co. Inc Supplier No. SE5027PK
Add to Cart
Add to Cart

Lima beans for use with a variety of applications

  • Accessory suitable for a wide range of planting and growing applications
  • Ideal for home gardens, educational use, and agricultural projects
  • Easy to handle and compatible with various soil types
  • Helps support versatile crop production and experimentation

Specifications

Quantity 1 Packet
Sufficient For Class of 15 students
Product Type Seed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.