Learn More
Invitrogen™ LIME Monoclonal Antibody (LIME-06)

Description
This antibody The antibody LIME-06 was raised against intracellular fragment corresponding to aa 141-295 of human LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed by T cells.
Specifications
Specifications
| Antigen | LIME |
| Applications | Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | LIME-06 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | Lime1 |
| Gene Accession No. | Q9H400 |
| Gene Alias | 2810038K19Rik; dJ583P15.4; FLJ20406; Lck interacting transmembrane adaptor 1; lck-interacting membrane protein; Lck-interacting molecule; lck-interacting transmembrane adapter 1; Lime; lime 1; LIME1; LP8067; RP4-583P15.5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.