Learn More
Grainger Limit Switch Body 1NO/1NC
Supplier: Grainger 14H177
The Dayton Limit Switch Body is a limit switch with 1 normally open and 1 normally closed contact. It has a NEMA 1 general purpose enclosure rating and is suitable for indoor use. The switch actuates when an actuator presses the lever. It has a maximum switching capacity of 10 Amps at 125/250V AC and a maximum operating force of 15 oz. The switch body mounts using (2) #6 or #8 screws and has 0.5 inch NPT conduit entries. It can be used in applications like conveyors, machine tools, transfer lines, packaging equipment, and more. The switch body is made of zinc alloy and has a black finish. It is UL/CSA certified.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.