Learn More
ECM Bioscience LIMK Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience LK6380
Encompass_Preferred
The LIMK phospho-regulation antibody sampler kit can be used to monitor the level of phosphorylated LIMK1 relative to total LIMK1 expression. In addition, the kit provides peptides for antibody blocking experiments. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.