Learn More
Invitrogen™ LIMS2 Polyclonal Antibody

Description
LIMS2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | LIMS2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Lims2 |
| Gene Accession No. | Q7Z4I7, Q91XD2 |
| Gene Alias | ILK-binding protein; LGMD2W; LIM and senescent cell antigen like domains 2; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM zinc finger domain containing 2; LIM-like protein 2; Lims2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; Pinch2; PINCH-2 |
| Gene Symbols | Lims2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.