Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIMS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142617
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142617 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142617 Supplier Invitrogen™ Supplier No. PA5142617
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 1 (NP_001129509.2) and isoformn 3 (NP_001154875.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

Adapter protein in a cytoplasmic complex linking beta- integrins to the actin cytoskeleton, bridges the complex to cell surface receptor tyrosine kinases and growth factor receptors. Plays a role in modulating cell spreading and migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Lims2
Gene Accession No. Q7Z4I7
Gene Alias ILK-binding protein; LGMD2W; LIM and senescent cell antigen like domains 2; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM zinc finger domain containing 2; LIM-like protein 2; Lims2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; Pinch2; PINCH-2
Gene Symbols Lims2
Host Species Goat
Immunogen Peptide with sequence C-RWSNMSDALA (aa 24-33).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55679
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.