Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN28A Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-7175190
Change view
Click to view available options
Quantity:
100 μg
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1096 100 μg
PIPA1096X 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA1096 Supplier Invitrogen™ Supplier No. PA1096
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIN28A Polyclonal Antibody for Flow, ICC/IF, IHC (P), IP, Western Blot

Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN28A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 1mg/mL BSA and 0.05% sodium azide
Gene LIN28A
Gene Accession No. Q8K3Y3, Q9H9Z2
Gene Alias AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; Lin28; LIN-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein LIN-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1
Gene Symbols LIN28A
Host Species Rabbit
Immunogen Full-length human recombinant protein expressed in bacteria.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79727, 83557
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.