Learn More
LIN28A Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | LIN28A |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | LIN28A |
| Gene Accession No. | Q8K3Y3, Q9H9Z2 |
| Gene Alias | AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; lin28; lin-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; Lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein lin-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; Testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1 |
| Gene Symbols | LIN28A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.