Learn More
Invitrogen™ LIN7A Monoclonal Antibody (P879)
Description
Recommended positive controls: MCF-7, MDA-MB-231. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | LIN7A |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | P879 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | LIN7A |
| Gene Accession No. | O14910 |
| Gene Alias | AI848705; hLin-7; lin 7 homolog a; LIN7; lin-7 homolog A (C. elegans); lin-7 homolog A, crumbs cell polarity complex component; Lin7a; lin-7A; lin-7-Ba; Mals1; MALS-1; mammalian LIN-7 1; mammalian lin-seven protein 1; mLin-7; protein lin-7 homolog A; Tax interaction protein 33; TIP-33; Veli; Veli1; veli-1; veli-1 protein; vertebrate homolog of C. elegans Lin-7 type 1; vertebrate LIN7 homolog 1; vertebrate lin-7 homolog 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.