Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN7B Monoclonal Antibody (OTI1B2), TrueMAB™, OriGene
SDP

Catalog No. p-200010424
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2286 30 μL
50-167-2579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2286 Supplier OriGene Supplier No. TA501435S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIN7B Monoclonal antibody specifically detects LIN7B in Human samples. It is validated for Western Blot

Specifications

Antigen LIN7B
Applications Western Blot
Classification Monoclonal
Clone OTI1B2
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FADD
Gene Accession No. Q13158
Gene Alias hLin7B; hVeli2; lin 7 homolog b; lin-7 homolog B; lin-7 homolog B (C. elegans); lin-7 homolog B, crumbs cell polarity complex component; lin-7-A; LIN7B; lin-7B; MALS2; MALS-2; Mammalian lin-seven protein 2; protein lin-7 homolog B; UNQ3116/PRO10200; Veli1a; Veli2; Veli-2; veli-2 protein; vertebrate homolog of C. elegans Lin-7 type 2; vertebrate lin-7 homolog 2
Host Species Mouse
Immunogen Full length human recombinant protein of human FADD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8772
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.