Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Linavonkibart Recombinant Monoclonal Antibody

Catalog No. MA558788
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA558788 1 mg
MA558787 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA558788 Supplier Invitrogen™ Supplier No. MA558788
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Linavonkibart, also known as SRK-181, is a fully human IgG4 monoclonal antibody designed to selectively inhibit the latent transforming growth factor-beta isoform 1 (TGFB1). Transforming growth factor-B1 (TGF-B1) is a key member of the TGF-beta family, a group of evolutionarily conserved polypeptide growth factors. The gene encoding TGF-B1 is located on chromosome 19 at the 19q13.2 locus. Structurally, TGF-B1 is synthesized as a precursor, which is then cleaved to produce a mature polypeptide that forms a dimeric protein. Functionally, TGF-B1 plays a crucial role in cellular processes such as proliferation, differentiation, and apoptosis, and is essential for regulating immune responses, maintaining tissue homeostasis, and orchestrating wound healing. Dysregulation of TGF-B1 signaling has been implicated in various pathological conditions including cancer, fibrosis, and autoimmune disorders. The activation of latent TGF-B1 stored in the extracellular matrix is mediated by interaction with integrins and other co-factors that induce conformational changes necessary for its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Linavonkibart
Applications ELISA, Functional Assay
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Host Species Human
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG4SP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.