Learn More
TOKU-E Lincomycin Hydrochloride, USP, 7179-49-9 , 5 g
Supplier:  TOKU-E L0025G
Encompass_Preferred
lincosamide antibiotic, C18H34N2O6S HCl H2O. Useful in microbiology, susceptibility testing, plant biology.
Catalog No. 50-255-1778