Learn More
Sigma Aldrich Fine Chemicals Biosciences Line position and line shape standard for powder diffraction NIST(R) SRM(R) 660c, Lanthanum hexaboride powder | 12008-21-8 | MFCD00151350 |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST660C
Encompass
Line position and line shape standard for powder diffraction NIST(R) SRM(R) 660c, Lanthanum hexaboride powder | Mol Wt: 203.77 | 12008-21-8 | MFCD00151350 |
Catalog No. 50-176-5870
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.