Learn More
JDM SCIENTIFIC LLC Linear barcode scanner
Supplier:  JDM SCIENTIFIC LLC 701113
Encompass
External handheld linear barcode scanner. High performance 1D linear scanner. Accurate first time scanning; always ready for the next scan. Decodes linear barcodes into any open application. Can be used alone, or with AlteRead and VolumeCheck 384 software, to capture rack and plate linear barcodes.
Catalog No. 50-271-9955