Learn More
Clontech Labs 3P LINEAR HYGROMYCIN MARKER
SDP
Supplier:  Clontech Labs 3P 631625
Encompass
Linear Hygromycin Marker, Size: 2microgram, it is a short ds DNA fragment consisting of the hygromycin resistance gene regulated by the SV40 promoter and ending with the SV40 polyadenylation signal. Preparation and Storage: At -20degreeC
Catalog No. NC9160477
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.