Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FLQ HbA1c for Abbott Systems
SDP

Catalog No. 22157984
Change view
Click to view available options
HbA1c
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-157-984 HbA1c
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-157-984 Supplier AUDIT™ MicroControls™ Supplier No. K949M5
This item is not returnable. View return policy

AUDIT™ Microcontrol™ Linearity FLQ HbA1c for Abbott Systems is assay quality control material consisting of five levels.

Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.

Specifications

Content And Storage <-15°C
Detectable Analytes HbA1c
For Use With (Equipment) Analyzers
Level A to E
Lyophilized No
Packaging Box
Product Name Linearity FLQ HbA1c for Abbott Systems
Stability 7 days when stored at 2 to 8°C
Form Frozen Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.