Learn More
Checkers Linebacker 1.35 in. Max Diameter Cable Protectors LB125 Checkers
Catalog No. 19088467 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Yellow, Orange
Catalog No. Color
19-088-467 Yellow, Orange
1 options
Catalog No. 19-088-467 Supplier Checkers Supplier No. CP5X125Y/O
Add to Cart
Add to Cart
Linebacker™ 5-channel cable protector with hinged lid provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage.
  • Protects cables and hoses up to 1.35 in. outside dia.
  • Patented T-Connectors to extend to any length
  • Reinforced hinged lid for easy cable placement
  • All-weather polyurethane construction
  • Patented 5-bar tread surface for maximum traction
  • Easy to transport, setup, disassemble, and store
  • Ideal for use in oil and gas applications, mining, military, and other industrial environments with heavy trucks and forklifts
Specifications
Color Yellow, Orange
Product Type Cable Protector
Material Polyurethane
Dimensions (L x W x H) 38.75 x 19.75 x 2.25 in.