Learn More
Checkers Linebacker 2.25 in. Max Diameter ADA Ramps LBRMP Checkers
Catalog No. 19088465 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
19-088-465 Blue
1 options
Catalog No. 19-088-465 Supplier Checkers Supplier No. CPRP1X225BLU
Only null left
Add to Cart
Add to Cart
Accessibility Ramps and Rails feature a gradual slope and slip-resistant surface to ensure a safer crossing point for carts, forklifts, vehicles with small wheels, and heavy trucks.
  • Patented, modular ADA/DDA Compliant ramps
  • Add ramps and rails when requirements specify
  • One person installation - no tools required
Specifications
Dimensions (L x W x H) 38.5 x 21.75 x 2.25 in.
Material Polyurethane
Product Type Cable Protector
Color Blue