Learn More
Checkers Linebacker 3 in. Max Diameter Cable Protectors LB300-GM Checkers
Catalog No. 19088468 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Orange, Yellow
Catalog No. Color
19-088-468 Orange, Yellow
1 options
Catalog No. 19-088-468 Supplier Checkers Supplier No. CP4X300Y/O
Add to Cart
Add to Cart
General Manufacturing Linebacker Cable Protector with hinged lid protects valuable electrical cables and hose lines from damage while providing a safer crossing in high volume traffic areas. Modular interlocking design is ideal for use in industrial manufacturing facilities and heavy equipment areas.
  • Protects Cables and Hoses Up to 3.0 in. Outside Dia.
  • Patented T-Connectors to Extend to Any Desired Length
  • Thick Lid and Base Allow for High Volume of Traffic
  • Reinforced Hinged Lid for Easy Cable Placement
  • Modular Interlocking Design
  • Ideal for Industrial Manufacturing Facilities and Heavy Equipment Areas
Specifications
Material Polyurethane
Dimensions (L x W x H) 40 x 20.25 x 6 in.
Color Orange, Yellow
Product Type Cable Protector