Learn More
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Catalog No. 03050282
Change view
Click to view available options
Description:
Graphite Liner Sealing Ring
Liner Sealing Ring
Viton Liner Sealing Ring
For Use With (Equipment):
Agilent Direct Straight Liners
Agilent SSL Liners
TRACE 1300 Series GC SSL Injectors
TRACE 1300 Series GC and Agilent SSL Injectors
TRACE and FOCUS GC SSL Injectors
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description For Use With (Equipment)
03-050-282 Viton Liner Sealing Ring TRACE 1300 Series GC and Agilent SSL Injectors
03-050-212 Liner Sealing Ring TRACE and FOCUS GC SSL Injectors
03-050-229 Graphite Liner Sealing Ring Agilent SSL Liners
01-189-303 Liner Sealing Ring TRACE 1300 Series GC SSL Injectors
03-050-228 Graphite Liner Sealing Ring Agilent Direct Straight Liners
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-050-282 Supplier Thermo Scientific™ Supplier No. 290OA241
Only null left
Add to Cart
Edge
Add to Cart
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viton Liner Sealing Ring
Quantity 10 Pack
Description Viton Liner Sealing Ring
For Use With (Equipment) TRACE 1300 Series GC and Agilent SSL Injectors