Learn More
R&D Systems™ LinkAble™ APC Antibody Conjugation Kits
SDP
Catalog No. RD10188113 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
3 x 100 μg
3 x 25 μg
5 x 1000 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10188113 25 μg
RD10188056 5 x 1000 μg
RD10188103 3 x 100 μg
RD10188104 3 x 25 μg
RD10188053 1 mg
RD10188055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RD10188113 Supplier R&D Systems™ Supplier No. LNKAPC25
Customers require a valid business account associated to their profile to access pricing and ordering options.
Supports reliable, reproducible conjugation with minimal hands-on time and no requirement for specialized expertise or additional purification steps.
Specifications
For Use With (Application) Flow Cytometry
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 25 μg