Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LinkAble™ PE Antibody Conjugation Kits
SDP

Catalog No. RD10188114 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
3 x 100 μg
3 x 25 μg
5 x 1000 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10188114 1 mg
RD10188097 100 μg
RD10188041 3 x 25 μg
RD10188042 5 x 1000 μg
RD10188063 3 x 100 μg
RD10188110 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RD10188114 Supplier R&D Systems™ Supplier No. LNKPE1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supports reliable, reproducible conjugation with minimal hands-on time and no requirement for specialized expertise or additional purification steps.

Specifications

For Use With (Application) Flow Cytometry
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Multi-species
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.