Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 38x6mm for Microplates, Assorted, 3900 labels, 25 sheets/Cs
Supplier: Alkali Scientific LCL14112A
CRYO LABEL SHEETS 38x6mm for Microplates We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.