Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linoleic Anhydride 90.0+%, TCI America™
SDP

Catalog No. L01451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01451G 1 g
L01455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01451G Supplier TCI America Supplier No. L01451G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24909-68-0
Molecular Formula C36H62O3
Molecular Weight (g/mol) 542.89
MDL Number MFCD00010456
InChI Key BDTYMGCNULYACO-MAZCIEHSSA-N
Synonym cis,cis-9,12-Octadecadienoic Anhydride
PubChem CID 15594594
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC

Specifications

Quantity 1 g
Formula Weight 542.89
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Linoleic Anhydride
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.