Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Linolenic acid, 99%

Catalog No. AC302820010
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302820010 1 g Glass bottle
AC302825000 500 mg Glass bottle
AC302820050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC302820010 Supplier Thermo Scientific Chemicals Supplier No. 302820010
  • $222.50
    $184.65 / Each of 1

    Save $37.85

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 463-40-1 | C18H30O2 | 278.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 463-40-1
Molecular Formula C18H30O2
Molecular Weight (g/mol) 278.44
MDL Number MFCD00065720
InChI Key DTOSIQBPPRVQHS-PDBXOOCHSA-N
Synonym linolenic acid, alpha-linolenic acid, linolenate, 9z,12z,15z-octadeca-9,12,15-trienoic acid, a-linolenic acid, cis,cis,cis-9,12,15-octadecatrienoic acid, all-cis-9,12,15-octadecatrienoic acid, alpha-linolenate, 9-cis,12-cis,15-cis-octadecatrienoic acid, z,z,z-9,12,15-octadecatrienoic acid
PubChem CID 5280934
ChEBI CHEBI:27432
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoic acid
SMILES CCC=CCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -11.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 230.0°C to 232.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2CH=CH)3(CH2)7CO2H
Refractive Index 1.48
Quantity 1 g
Beilstein 02, IV, 1781
Specific Gravity 0.91
Merck Index 15, 5561
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 278.44
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Linolenic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-334-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.