Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Linseed oil

Catalog No. 4300211L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-002-11L 1 L
43-002-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-002-11L Supplier Sigma Aldrich Supplier No. 4300211L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Linseed oil is a natural triglyceride and a well known drying oil. Drying of the triglyceride mainly leads to the oxidation of the unsaturated fatty acid segments and polymerization. Oxidative degradation studies of linseed oil show hardening and oxidation of alkylic sections followed by partial fragmentation of the triglyceride structure.Linseed oil filled urea-formaldehyde resin microcapsules may be used to fix cracks in paints/coatings.

Specifications

CAS 8001-26-1
Density 0.93 g/mL (at 25°C)
Refractive Index n20/D 1.4795 (literature)
Quantity 1 L
MDL Number MFCD00131507
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.