Learn More
Cayman Chemical Lipase Activity Assay Kit, 96 wells
Supplier: Cayman Chemical 70064096WELLS
Lipases perform essential roles in the digestion, transportation, and processing of dietary lipids by controlling the clearance of triglyceride-rich lipoproteins from the circulation. Manipulating lipolysis has therapeutic potential in the metabolic disorders frequently associated with obesity. Cayman’s Lipase Activity Assay provides a fluorescence-based method for detecting lipase activity in plasma, serum, tissue homogenates, and cell culture samples. In the assay, lipase hydrolyzes arachidonoyl-1-thioglycerol to arachidonic acid and thioglycerol. Thioglycerol reacts with the thiol fluorometric detector to yield a highly fluorescent product which can be analyzed with an excitation wavelength of 380-390 nm and an emission wavelength of 510-520 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.