Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lipase, MP Biomedicals™

Catalog No. MP210218920 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210218920 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210218920 Supplier MP Biomedicals™ Supplier No. 0210218920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catalyze the hydrolysis and synthesis of the emulsified esters of glycerol and long chain fatty acids. Lipase, MP Biomedicals is a subclass of the esterases. It is used industrially for the resolution of chiral compounds and the trans-esterification produ

Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions.They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).

TRUSTED_SUSTAINABILITY

Specifications

Color Cream
For Use With (Application) Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Content And Storage -20°C
Source Porcine
Quantity 20 g
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.