Learn More
Thermo Scientific Chemicals Lipase, from porcine pancreas
Catalog No. AAJ6290336
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
AAJ6290336 500 g
1 options
Catalog No. AAJ6290336 Supplier Thermo Scientific Chemicals Supplier No. J6290336
Add to Cart
Lipase, is used as a catalyst in the preparation of biodiesels and in the preparation of esters of chiral epoxy alcohols. Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 g
Form Powder