Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lipase Powder
SCI_ED

Catalog No. S25731 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25731 25 g
S25731A 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25731
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triacylglycerol Hydrolase; CAS: 9001-62-1

Assay Percent Range: >99% w/v
Soluble

This product is for educational use - no C of A is available.

Specifications

Color Beige
Purity >99%
For Use With (Application) Laboratory chemicals
Quantity 25 g
Product Type Lipase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.